Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Antibody (85B152.5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197728PCP
Description
TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular).Specifications
TLR5 | |
Monoclonal | |
PerCP | |
O60602 | |
TLR5 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259). | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Canine | |
Purified |
Flow Cytometry | |
85B152.5 | |
Flow Cytometry 1 μL/1 million cells, Flow (Cell Surface), Flow (Intracellular) | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
Mouse | |
100 kDa | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
7100 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction