Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLR5 Monoclonal Antibody (8-1#)

Catalog No. PIMA543703
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543703 20 μL
PIMA541682 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543703 Supplier Invitrogen™ Supplier No. MA543703
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR5 Monoclonal Antibody for Western Blot, IHC (P)

TLR5 is a Type I membrane protein belonging to the Toll-like receptor family, participates in the innate immune response to microbial agents. TLR5 also plays a role in mediating detection of bacterial flagellins. TLR5 acts via MyD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response. TLR5 protein binds to TIRAP and MyD88 via their respective TIR domains TLR5 is highly expressed in liver, and is detected in lung and at very low levels in most other tissues. The TLR5 gene lies in a locus that is associated with susceptibility to Salmonella. Inbred strains of mice can be classified into 3 categories according to their resistance to infection with S.typhimurium: susceptible (BALB/c, C57BL/6, C3H/He), intermediate (DBA/2, C75L) and resistant (A, CBA). The strain MOLF/Ei is highly susceptible to the infection, has a unique TLR5 haplotype and a lower expression of TRL5. A common TLR5 stop codon polymorphism that disrupts TLR5 signaling is associated with susceptibility to Legionnaires' disease and demonstrates the importance of TLR5 in the innate immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8-1#
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TLR5
Gene Accession No. O60602
Gene Alias FLJ10052; MELIOS; MGC126430; MGC126431; RP11-239E10.1; SLE1; SLEB1; systemic lupus erythematosus susceptibility 1; TIL3; Tlr5; toll like receptor 5; Toll/interleukin-1 receptor-like protein 3; toll-like receptor 5
Gene Symbols TLR5
Host Species Mouse
Immunogen Recombinant protein Toll Like Receptor 5. The antigen corresponds to amino acid range 693-858 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289337, 7100
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.