Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Polyclonal Antibody
TLR5 Polyclonal Antibody
Supplier: Thermo Scientific PA595257
Description
TLR5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow

Specifications
TLR5 | |
Polyclonal | |
Unconjugated | |
TLR5 | |
FLJ10052; MELIOS; MGC126430; MGC126431; RP11-239E10.1; SLE1; SLEB1; systemic lupus erythematosus susceptibility 1; TIL3; Tlr5; toll like receptor 5; Toll/interleukin-1 receptor-like protein 3; toll-like receptor 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7100 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O60602 | |
TLR5 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human TLR5 (801-828aa MKHQSIRGFVQKQQYLRWPEDFQDVGWF). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction