Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Rabbit anti-Human, Mouse, Clone: 6P1Q10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | TLR5 |
---|---|
Clone | 6P1Q10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TLR5 Monoclonal antibody specifically detects TLR5 in Human, Mouse samples. It is validated for Western BlotSpecifications
TLR5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7100 | |
IgG | |
Affinity purified |
6P1Q10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 258-404 of human TLR5 (O60602). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title