Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR6 Antibody (418601) [DyLight 755], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1533Z
Description
TLR6 Monoclonal antibody specifically detects TLR6 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
TLR6 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
418601 | |
Flow Cytometry, CyTOF-ready | |
CD286, CD286 antigen, toll-like receptor 6 | |
HEK293 human embryonic kidney cell line transfected with mouse TLR6, Phe39-Thr806, Accession # BAA78632 | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
10333 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction