Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR6 Antibody (86B1153.2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056536AF750
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).Specifications
TLR6 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TLR6 | |
This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. | |
0.1 mL | |
Primary | |
TLR6 (86B1153.2) | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
86B1153.2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular) | |
CD286, CD286 antigen, toll-like receptor 6 | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
10333.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction