Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR6 Antibody (86B1153.2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056536IR
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).Specifications
TLR6 | |
Monoclonal | |
DyLight 755 | |
Q9Y2C9 | |
TLR6 | |
This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
10333 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
86B1153.2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular) | |
CD286, CD286 antigen, toll-like receptor 6 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
TLR6 (86B1153.2) | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction