Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR6 (CD286) Mouse anti-Human, PE, Clone: TLR6.127, BD

Catalog No. BDB566339 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566339 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566339 Supplier BD Biosciences Supplier No. 566339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TLR6.127 monoclonal antibody specifically recognizes Toll-like receptor 6 (TLR6), which is also known as CD286. CD286 belongs to the mammalian Toll-like receptor (TLR) family of pattern recognition receptors that participate in inducing innate and adaptive immune responses to microbial pathogens. TLR6 is a type I transmembrane glycoprotein comprised of an extracellular domain with a large number of leucine?rich repeats (LRRs). Its cytoplasmic region contains a signaling Toll/Interleukin-1 Receptor (TIR) domain. TLR6 is expressed on monocytes, neutrophils, and some dendritic cells. In association with TLR2 (CD282), TLR6 recognizes diacylated lipopeptides and peptidoglycans of bacterial or fungal origin. The ligand-bound TLR2:TLR6 complex transduces responses intracellularly through MyD88 and the NF- B signaling cascade leading to the production of proinflammatory cytokines by responding cells.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR6 (CD286)
Classification Monoclonal
Clone TLR6.127
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Toll-like receptor 6, CD286
Host Species Mouse
Immunogen Human TLR6 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.