Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLR6 Monoclonal Antibody (86B1153.2), PE
GREENER_CHOICE

Catalog No. PIMA516177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516177 Supplier Invitrogen™ Supplier No. MA516177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cross reacts with Human.

TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR6
Applications Flow Cytometry
Classification Monoclonal
Clone 86B1153.2
Concentration 0.51 mg/mL
Conjugate PE
Formulation PBS with 0.05% sodium azide
Gene TLR6
Gene Accession No. Q9Y2C9
Gene Alias CD antigen 286; CD286; TLR6; toll like receptor 6; toll-like receptor 6
Gene Symbols TLR6
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 408-424 of human TRL6.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10333
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.