Learn More
Invitrogen™ TLR6 Monoclonal Antibody (86B1153.2)

Description
Suggested positive control: antigen standard for TLR6 (transient overexpression lysate), Ramos.
Specifications
Specifications
| Antigen | TLR6 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 86B1153.2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TLR6 |
| Gene Accession No. | Q9Y2C9 |
| Gene Alias | CD antigen 286; CD286; TLR6; toll like receptor 6; toll-like receptor 6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.