Learn More
TLR7 Antibody (4G6), Alexa Fluor™ 532, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP225274AF532
Catalog No. NP22274A32
Only null left
Add to Cart
Description
Description
TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
Specifications
TLR7
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
TLR7
A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen.
Primary
Human, Mouse
IgG1 κ
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, KnockDown
4G6
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, KnockDown Validated
toll-like receptor 7
Mouse
0.1 mL
51284
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.