Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR7 Antibody (4G6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225274APC
Description
TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Knockdown Validated.Specifications
TLR7 | |
Monoclonal | |
APC | |
Q9NYk1 | |
TLR7 | |
A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
51284 | |
Human, Mouse | |
Purified |
Flow Cytometry, KnockDown | |
4G6 | |
Flow Cytometry, KnockDown Validated | |
toll-like receptor 7 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human. Mouse reactivity reported in scientific literature (PMID: 24126361). | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction