Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (4G6) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 4 publications

$268.00 - $548.00

Specifications

Antigen TLR7
Clone 4G6
Concentration 1.0 mg/mL
Dilution Western Blot 10 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Immunohistochemistry-Frozen reported in scientific literature (PMID 22610069), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22610069), CyTOF-ready, KnockDown Validated
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF, KnockDown
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225274SS
SDP
View Documents
Novus Biologicals
NBP225274SS
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP225274
SDP
View Documents
Novus Biologicals
NBP225274
0.1 mg
Each for $548.00
Only null left
Add to Cart
 
Description

Description

TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, CyTOF-ready, Knockdown Validated.
Specifications

Specifications

TLR7
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF, KnockDown
Unconjugated
Mouse
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
NP_057646
51284
A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4G6
Western Blot 10 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Immunohistochemistry-Frozen reported in scientific literature (PMID 22610069), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22610069), CyTOF-ready, KnockDown Validated
Monoclonal
Purified
RUO
Human, Mouse
toll-like receptor 7
TLR7
IgG1 κ
Protein G purified
This product reacts with Human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.