Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR7 Antibody (533707), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5875MFV500
Description
TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| TLR7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 51284 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 533707 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| toll-like receptor 7 | |
| E. coli-derived recombinant human TLR7, Met360-Leu516, Accession # Q9NYK1 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction