Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR7 Antibody (533707), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5875C
Description
TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
TLR7 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
533707 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
toll-like receptor 7 | |
E. coli-derived recombinant human TLR7, Met360-Leu516, Accession # Q9NYK1 | |
0.1 mL | |
Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
51284 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction