Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR7 Antibody (6E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $479.73
Specifications
| Antigen | TLR7 |
|---|---|
| Clone | 6E5 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
Description
TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| TLR7 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| toll-like receptor 7 | |
| A synthesized peptide derived from Human TLR7 [UniProt Q9NYK1] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6E5 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 51284 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title