Learn More
Invitrogen™ TLR7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595046
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Spleen Tissue, Rat Liver Tissue, U87 Cell, A549 Cell. IHC: Human Placenta Tissue, Rat Spleen Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TLR7 is a 10 amino acid long highly conserved membrane protein belonging to the Toll-like receptor (TLR) family with 27 LRR (leucine-rich) repeats and a TIR domain. TLR7 plays a fundamental role in pathogen recognition and activation of innate immunity. TLR7 is activated by infections with single-stranded RNA viruses, including influenza virus and vesicular stomatitis virus (VSV). TLR7 is predominantly expressed in lung, placenta, and spleen, and lies in close proximity to another family member, TLR8. TLR7 also acts via MyD88 and TRAF6, leading to NF-kappaB activation, cytokine secretion and the inflammatory response. TLR7 recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity.
Specifications
TLR7 | |
Polyclonal | |
Unconjugated | |
TLR7 | |
RGD1563357; RP23-139P21.3; Tlr7; TLR7-like; toll like receptor 7; toll-like receptor 7; toll-like receptor 7-like; UNQ248/PRO285 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
170743, 317468, 51284 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P58681, Q9NYK1 | |
TLR7 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human TLR7 (1020-1038aa QAHPYFWQCLKNALATDNH). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.