Learn More
Invitrogen™ TLR7 Polyclonal Antibody, FITC

Description
This antibody will cross-react with mouse TLR7. In human Ramos cells, additional bands are seen.
Specifications
Specifications
| Antigen | TLR7 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.05 mg/mL |
| Conjugate | FITC |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | TLR7 |
| Gene Accession No. | P58681, Q9NYK1 |
| Gene Alias | RGD1563357; RP23-139P21.3; Tlr7; TLR7-like; toll like receptor 7; toll-like receptor 7; toll-like receptor 7-like; UNQ248/PRO285 |
| Gene Symbols | TLR7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.