Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR8 Antibody (44C143), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224917AF488
Description
TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
TLR8 | |
Monoclonal | |
Alexa Fluor 488 | |
Q9NR97 | |
TLR8 | |
This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850. | |
0.1 mL | |
Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
51311 | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
44C143 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody is reactive to Human, Mouse. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction