Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR8 Antibody (44C143), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224917MFV610
Description
TLR8 Monoclonal antibody specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)Specifications
TLR8 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rabbit | |
Purified |
Flow Cytometry | |
44C143 | |
Flow Cytometry, Flow (Cell Surface), Flow (Intracellular) | |
CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850. | |
0.1 mL | |
Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
51311 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction