Learn More
TLR8 Mouse anti-Human, CoraLite 594, Clone: 3B6A4, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL59467317100UL
Description
The TLR8 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TLR8. This antibody recognizes human antigen. The TLR8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
TLR8 | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
51311 | |
IgG1 |
Immunofluorescence | |
3B6A4 | |
Mouse | |
Primary | |
Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.