Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Antibody (1138D) [DyLight 405], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB7960E
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), ImmunocytochemistrySpecifications
TLR9 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
1138D | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
CD289, CD289 antigen, toll-like receptor 9 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9, Leu26-Asp818, Accession # AAK29625 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
54106 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction