Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    TLR9 Antibody (1138D), mFluor Violet 610 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB7960MFV610
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| TLR9 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 54106 | |
| Store at 4C in the dark. | 
| Flow Cytometry, Immunocytochemistry | |
| 1138D | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9, Leu26-Asp818, Accession # AAK29625 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            