Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Antibody (26C593R), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB36583P
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
TLR9 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
26C593R | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
CD289, CD289 antigen, toll-like receptor 9 | |
Antibody developed against the KLH-conjugated synthetic peptide corresponding to amino acids Asp250-Asn300 of human TLR9 isoform A., Accession # Q9NR96 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
54106 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction