Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Antibody (eB72-1665), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143140G
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TLR9 | |
Monoclonal | |
DyLight 488 | |
Q9NR96 | |
TLR9 | |
The immunogen for this antibody was Tolllike receptor 9. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
54106 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
eB72-1665 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD289, CD289 antigen, toll-like receptor 9 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction