Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176680PCP
Description
TLR9 Polyclonal antibody specifically detects TLR9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
TLR9 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
CD289, CD289 antigen, toll-like receptor 9 | |
Antibody was raised against a peptide corresponding to 15 amino acids near the center of human TLR9. The immunogen is located within amino acids 840 - 890 of TLR9. Amino Acid Squence: RGRQSGRDEDALPYD | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
54106 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction