Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Antibody (SR1133), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TLR9 |
---|---|
Clone | SR1133 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Western BlotSpecifications
TLR9 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD289, CD289 antigen, toll-like receptor 9 | |
A synthesized peptide derived from human TLR9 (Uniprot #: Q9NR96) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1133 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
54106 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title