Learn More
Invitrogen™ TLR9 Monoclonal Antibody (5G5), Biotin
Description
The monoclonal antibody 5G5 reacts with RAW macrophages and TLR9 transfected HEK293 cells, and is cross reactive with canine TLR9.
Specifications
Specifications
| Antigen | TLR9 |
| Applications | Inhibition Assays, Western Blot |
| Classification | Monoclonal |
| Clone | 5G5 |
| Concentration | 100 μg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | Tlr9 |
| Gene Accession No. | Q9EQU3, Q9NR96 |
| Gene Alias | CD289; Tlr9; toll like receptor 9; toll-like receptor 9; UNQ5798/PRO19605 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.