Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TLR9 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC3658R100UG
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| TLR9 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_059138 | |
| TLR9 | |
| NS0 mouse myeloma cell line transfected with human TLR9 Accession # NP_059138 | |
| RUO | |
| 54106 | |
| Human | |
| IgM |
| Flow Cytometry | |
| 229106 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TLR9. Stains human TLR9-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction