Learn More
Invitrogen™ TLR9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589960
Description
Positive Samples: Mouse spleen, Rat spleen; Cellular Location: Cytoplasmic vesicle, Endoplasmic reticulum membrane, Endosome, Lysosome, Single-pass type I membrane protein, phagosome Immunogen sequence: FPGLRYVDLS DNRISGASEL TATMGEADGG EKVWLQPGDL APAPVDTPSS EDFRPNCSTL NFTLDLSRNN LVTVQPEMFA QLSHLQCLRL SHNCISQAVN GSQFLPLTGL QVLDLSHNKL DLYHEHSFTE.
The protein encoded by this gene is a member of the Toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is preferentially expressed in immune cell rich tissues, such as spleen, lymph node, bone marrow and peripheral blood leukocytes. Studies in mice and human indicate that this receptor mediates cellular response to unmethylated CpG dinucleotides in bacterial DNA to mount an innate immune response.
Specifications
TLR9 | |
Polyclonal | |
Unconjugated | |
Tlr9 | |
CD289; Tlr9; toll like receptor 9; toll-like receptor 9; UNQ5798/PRO19605 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
338457, 54106, 81897 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9EQU3, Q9NR96 | |
Tlr9 | |
A synthetic peptide corresponding to a sequence within amino acids 414-543 of human TLR9 (NP_0591381). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.