Learn More
Invitrogen™ TLX Monoclonal Antibody (H6510)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ 434500
Description
This antibody specifically recognizes human TLX and cross reacts with mouse and rat TLX.
Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
Specifications
| TLX | |
| Monoclonal | |
| 1 mg/mL | |
| Physiological saline with 0.1% sodium azide | |
| Q64104, Q9Y466 | |
| NR2E1 | |
| Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant human TLX (81-180 aa). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| H6510 | |
| Unconjugated | |
| NR2E1 | |
| fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 21907, 684085, 7101 | |
| Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.