Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534003
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534003 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534003 Supplier Invitrogen™ Supplier No. PA534003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLX Polyclonal Antibody for IHC (P)

Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR2E1
Gene Accession No. Q9Y466
Gene Alias fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991
Gene Symbols NR2E1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminal extracellular domain of human NR2E1.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100072159, 100155153, 100328698, 475017, 528156, 7101
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.