Learn More
Invitrogen™ TLX Polyclonal Antibody

Description
Immunogen sequence: YFRGHKEENG AAAHFPSAAL PAPAFFTAVT QLEPHGLELA AVSTTPERQT LVSLAQPTPK YPHEVNGTPM YLYEVATESV CESAARLLFM SIKWAKSVPA FSTLSLQDQL MLLEDAWREL FVLGIAQWAI PVDANTLLAV SGMNGDNTDS QKLNKIISEI QALQEVVARF RQLRLDATEF ACLKCIVTFK AVPTHSGSEL RSFRNAAAIA ALQDEAQLTL NSYIHTRYPT QPCRFGKLLL LLPALRSISP STIEEVFFKK TIGNVPITRL LSDMYKSSDI; Positive Samples: HL-60, HepG2; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | TLX |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.93 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NR2E1 |
| Gene Accession No. | Q9Y466 |
| Gene Alias | fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991 |
| Gene Symbols | NR2E1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.