Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596684 Supplier Invitrogen™ Supplier No. PA596684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YFRGHKEENG AAAHFPSAAL PAPAFFTAVT QLEPHGLELA AVSTTPERQT LVSLAQPTPK YPHEVNGTPM YLYEVATESV CESAARLLFM SIKWAKSVPA FSTLSLQDQL MLLEDAWREL FVLGIAQWAI PVDANTLLAV SGMNGDNTDS QKLNKIISEI QALQEVVARF RQLRLDATEF ACLKCIVTFK AVPTHSGSEL RSFRNAAAIA ALQDEAQLTL NSYIHTRYPT QPCRFGKLLL LLPALRSISP STIEEVFFKK TIGNVPITRL LSDMYKSSDI; Positive Samples: HL-60, HepG2, U-251MG, Jurkat, Mouse liver; Cellular Location: Nucleus.

Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NR2E1
Gene Accession No. Q64104, Q9Y466
Gene Alias fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991
Gene Symbols NR2E1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 106-385 of human NR2E1/TLX (NP_0032601).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21907, 7101
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.