Learn More
Invitrogen™ TLX2 Polyclonal Antibody

Description
Immunogen sequence: MEPGMLGPHNL PHHEPISFG Highest antigen sequence identity to the following orthologs - mouse 75%, rat 80%.
Specifications
Specifications
| Antigen | TLX2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TLX2 |
| Gene Accession No. | O43763 |
| Gene Alias | Enteric neuron homeobox protein; Enx; homeo box 11-like 1; homeobox 11-like1; homeobox protein Hox-11L1; Homeobox TLX-2; hox11l; Hox11L.1; Hox11l1; NCX; Ncx1; neural crest homeobox protein; PMUR10F; T cell leukemia homeobox 2; T cell leukemia, homeobox 2; T-cell leukemia associated homeodomain protein; T-cell leukemia associated homeodomain protein Tlx3a; T-cell leukemia homeobox 2; T-cell leukemia homeobox protein 2; T-cell leukemia, homeobox 2; T-cell leukemia, homeobox 3a; Tlx1l1; Tlx1l2; Tlx2; tlx3a; Tlx-3a; tlxA; transcription factor |
| Gene Symbols | TLX2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.