Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534554
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534554 Supplier Invitrogen™ Supplier No. PA534554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-34554 can be used with blocking peptide PEP-1597.

The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX2, also known as homeobox11-like 1 (HOX11L1), is a DNA-binding nuclear transcription factor that appears to be crucial for the development of the peripheral nervous system. TLX2 is a downstream target of regulation by both PHOX2A and PHOX2B proteins during intestinal neuronal differentiation. In humans, loss-of-function of TLX2 may play a role in tumorigenesis of gastrointestinal stromal tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TLX2
Gene Accession No. O43763, Q61663
Gene Alias Enteric neuron homeobox protein; Enx; homeo box 11-like 1; homeobox 11-like1; homeobox protein Hox-11L1; Homeobox TLX-2; hox11l; Hox11L.1; Hox11l1; NCX; Ncx1; neural crest homeobox protein; PMUR10F; T cell leukemia homeobox 2; T cell leukemia, homeobox 2; T-cell leukemia associated homeodomain protein; T-cell leukemia associated homeodomain protein Tlx3a; T-cell leukemia homeobox 2; T-cell leukemia homeobox protein 2; T-cell leukemia, homeobox 2; T-cell leukemia, homeobox 3a; Tlx1l1; Tlx1l2; Tlx2; tlx3a; Tlx-3a; tlxA; transcription factor
Gene Symbols TLX2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human TLX2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21909, 3196, 680117
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.