Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLX3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987147
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7147 Supplier Bioss Supplier No. BS6240R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX3, also known as homeobox11-like1 (HOX11L2), is a DNA-binding nuclear transcription factor that is expressed in a subset of the primary sensory nervous system. TLX3, along with the related TLX1, is a selector transcription factor that promotes an excitatory glutamatergic neuronal phenotype over an inhibitory GABAergic phenotype opposing LBX1 signals during dorsal spinal cord development. Chromosomal translocations of the TLX3 gene have been shown to result in some forms of T-cell acute lymphoblastic leukemia (T-ALL).

Specifications

Antigen TLX3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TLX3
Gene Accession No. O43711
Gene Alias homeo box 11-like 2; homeobox 11-like 2; Homeobox protein Hox-11L2; Homeobox TLX-3; Hox11l2; Respiratory neuron homeobox protein; RGD1564190; Rnx; T cell leukemia, homeobox 3; T-cell leukemia homeobox 3; T-cell leukemia homeobox protein 3; T-cell leukemia, homeobox 3; Tlx1l2; Tlx3
Gene Symbols TLX3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TLX3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30012
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.