Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TM 5275 sodium salt
GREENER_CHOICE

Catalog No. 576910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-691-0 10 mg
57-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-691-0 Supplier Tocris Bioscience™ Supplier No. 5769/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable

TM 5275 sodium salt is a plasminogen activator inhibitor-1 (PAI-1) inhibitor. Orally bioavailable.

Specifications

Chemical Name or Material 5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt
CAS 1103926-82-4
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C28H27ClN3NaO5
Formula Weight 543.97
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.