Learn More
Tocris Bioscience™ TM 5441

Description
Specifications
Specifications
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Formula Weight | 450.8 |
| Physical Form | Solid |
| Purity | >98% |
| Chemical Name or Material | 5-Chloro-2-[[2-[2-[[3-(3-furanyl)phenyl]amino]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt |
Safety and Handling
| Recommended Storage | Store at +4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.