Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM111 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559959 Supplier Invitrogen™ Supplier No. PA559959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVSDSQVLIR SRVLRENGKY IPKQSFLTRK YYFNNPEDGF FKKTKRKVVP PSPMTDPTM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TMEM111,also names as EMC3, is part of the endoplasmic reticulum-associated secretory pathway (PMID: 19797678). TMEM111 was required for murine pulmonary surfactant synthesis and lung function at birth (PMID: 29083321). It can be detect a band of 30 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM111
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EMC3
Gene Accession No. Q9P0I2
Gene Alias Transmembrane protein 111
Gene Symbols EMC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TM111. Recombinant protein control fragment (Product #RP-90947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55831
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.