Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM2D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570491
Description
This target displays homology in the following species: Cow: 92%; Dog: 93%; Goat: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. This gene has multiple alternatively spliced transcript variants which encode two different isoforms.
Specifications
TM2D2 | |
Polyclonal | |
Unconjugated | |
TM2D2 | |
2410018G23Rik; BBP-like protein 1; Beta-amyloid-binding protein-like protein 1; BLP1; RGD1306769; si:dkey-49o11.3; TM2 domain containing 2; TM2 domain containing 2, isoform a; TM2 domain-containing protein 2; TM2 domain-containing protein 2-like; TM2 domain-containing protein 2-like protein; TM2D2; Unknown (protein for MGC:133974); zgc:92201 | |
Rabbit | |
Affinity chromatography | |
RUO | |
83877 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BX73 | |
TM2D2 | |
synthetic peptide directed towards the middle region of human TM2D2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction