Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM2D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112283 Supplier Invitrogen™ Supplier No. PA5112283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TM2D2 Polyclonal Antibody for Western Blot

The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. This gene has multiple alternatively spliced transcript variants which encode two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM2D2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene TM2D2
Gene Accession No. Q9BX73
Gene Alias 2410018G23Rik; BBP-like protein 1; Beta-amyloid-binding protein-like protein 1; BLP1; RGD1306769; si:dkey-49o11.3; TM2 domain containing 2; TM2 domain containing 2, isoform a; TM2 domain-containing protein 2; TM2 domain-containing protein 2-like; TM2 domain-containing protein 2-like protein; TM2D2; Unknown (protein for MGC:133974); zgc:92201
Gene Symbols TM2D2
Host Species Rabbit
Immunogen E. coli-derived Human TM2D2 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83877
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.