Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM4SF18 Antibody (211116), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3509PEATT594
Description
TM4SF18 Monoclonal antibody specifically detects TM4SF18 in Human samples. It is validated for Flow CytometrySpecifications
TM4SF18 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
116441 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Flow Cytometry | |
211116 | |
Flow Cytometry | |
L6D | |
NS0 mouse myeloma cell line transfected with human TM4SF18, Met1-Ile201, Accession # Q96CE8 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction