Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM4SF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567048 Supplier Invitrogen™ Supplier No. PA567048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGQTSYASSNK LTNYVWYFE Highest antigen sequence identity to the following orthologs - mouse 50%, rat 50%.

Transmembrane 4 L Six Family Member 18 (TM4SF18) is a gene located on chromosome 11q12.3 that belongs to the transmembrane 4 L six superfamily, a group of proteins characterized by the presence of four transmembrane domains. TM4SF18 is expressed in various tissues and is involved in cellular processes such as cell adhesion, migration, and proliferation. As part of the tetraspanin family, TM4SF18 proteins play a role in organizing cellular membranes and facilitating interactions between cell surface receptors, impacting signaling pathways crucial for maintaining cellular functions and tissue homeostasis. Although TM4SF18's specific biological roles are still being elucidated, it has been implicated in tumorigenesis, where altered expression may influence cancer progression by modulating cell-cell communication and migration. Research into TM4SF18 aims to uncover its precise functions in health and disease, particularly its potential involvement in cancer biology and as a target for therapeutic interventions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM4SF18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TM4SF18
Gene Accession No. Q96CE8
Gene Alias AAB5; L6D; TM4SF18; transmembrane 4 L six family member 18; transmembrane 4 L6 family member 18
Gene Symbols TM4SF18
Host Species Rabbit
Immunogen Recombinant Human TM4SF18. Recombinant protein control fragment (Product #RP-107690).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.