Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF20 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15984820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15984820 20 μL
NBP159848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15984820 Supplier Novus Biologicals Supplier No. NBP15984820UL

Rabbit Polyclonal Antibody

TM4SF20 Polyclonal specifically detects TM4SF20 in Human samples. It is validated for Western Blot.

Specifications

Antigen TM4SF20
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q53R12
Gene Alias FLJ22800, PRO994, TCCE518, transmembrane 4 L six family member 20, transmembrane 4 L6 family member 20
Gene Symbols TM4SF20
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TM4SF20 (transmembrane 4 L six family member 20) The peptide sequence was selected from the middle region of TM4SF20)(50ug). Peptide sequence QALLKGPLMCNSPSNSNANCEFSLKNISDIHPESFNLQWFFNDSCAPPTG.
Molecular Weight of Antigen 25 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79853
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.