Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM4SF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107037
Description
Sequence homology: Cow: 79%; Guinea Pig: 79%; Human: 100% Immunogen sequence: CTGGCARCLG GTLIPLAFFG FLANILLFFP GGKVIDDNDH LSQEIWFFGG.
TM4SF4 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This protein is a cell surface glycoprotein that can regulate cell proliferation.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that can regulate cell proliferation. The use of alternate polyadenylation sites has been found for this gene.
Specifications
| TM4SF4 | |
| Polyclonal | |
| Unconjugated | |
| TM4SF4 | |
| Iltmp; IL-TMP; intestinal and liver (il) tetraspan membrane protein; intestine and liver tetraspan membrane protein; TM4SF4; transmembrane 4 L six family member 4; transmembrane 4 L6 family member 4; transmembrane 4 superfamily member 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7104 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P48230 | |
| TM4SF4 | |
| Synthetic peptide directed towards the N terminal region of human TM4SF4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction