Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM6SF1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548742

Catalog No. PA548742


Only null left
Add to Cart

Description

Description

Predicted to react with bovine based on sequence homology.

May function as sterol isomerase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TM6SF1
Polyclonal
Unconjugated
TM6SF1
TM6SF1; Transmembrane 6 superfamily member 1
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
53346
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9BZW5
TM6SF1
KLH conjugated synthetic peptide between 40-68 amino acids from the N-terminal region of human TM6SF1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.