Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM7SF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542735
Description
Peptide sequence: LAARSGPARL LGPPASLPGL EVLWSPRALL LWLAWLGLQA ALYLLPARKV Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 85%; Rabbit: 100%; Rat: 100%.
TM7SF2 is involved in the conversion of lanosterol to cholesterol.
Specifications
TM7SF2 | |
Polyclonal | |
Unconjugated | |
TM7SF2 | |
3110041O18Rik; 3-beta-hydroxysterol Delta (14)-reductase; ANG1; Another new gene 1 protein; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase; delta-14-SR; Dhcr14; DHCR14A; NET47; Putative sterol reductase SR-1; sterol C14-reductase; Tm7sf2; Transmembrane 7 superfamily member 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7108 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O76062 | |
TM7SF2 | |
Synthetic peptide directed towards the N-terminal of human TM7SF2 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction