Learn More
Invitrogen™ TM7SF2 Polyclonal Antibody

Description
Immunogen sequence: LAPGGNSGNP IYDFFLGREL NPRICFFDFK YFCELRPGLI GWVLINLALL MKEAELRGSP SLAMWLVNGF QLLYVGDALW HEEAVLTTMD I.
Specifications
Specifications
| Antigen | TM7SF2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TM7SF2 |
| Gene Accession No. | O76062, Q71KT5 |
| Gene Alias | 3110041O18Rik; 3-beta-hydroxysterol Delta (14)-reductase; ANG1; Another new gene 1 protein; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase; delta-14-SR; Dhcr14; DHCR14A; NET47; Putative sterol reductase SR-1; sterol C14-reductase; Tm7sf2; Transmembrane 7 superfamily member 2 |
| Gene Symbols | TM7SF2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.