Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM7SF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144053
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144053 Supplier Invitrogen™ Supplier No. PA5144053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human RT4 whole cell, human Jurkat whole cell, human MOLT-4 whole cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The TM7SF3 gene encodes transmembrane 7 superfamily member 3, a protein that is involved in cellular homeostasis and stress responses. TM7SF3 is characterized by its seven transmembrane domains, which contribute to its role in signal transduction across the cell membrane. This gene is expressed in various tissues and has been implicated in regulating cellular growth and survival pathways, proving critical in the context of cellular response to endoplasmic reticulum (ER) stress and maintaining cellular integrity. TM7SF3 modulates processes such as apoptosis and autophagy, particularly under stress conditions, thus influencing both normal physiological processes and pathological states like cancer. Research has shown that TM7SF3 expression can be altered in certain diseases, affecting pathways that govern cell cycle progression and stress adaptation. Understanding the role of TM7SF3 in cellular signaling and stress management can aid in identifying therapeutic targets for conditions characterized by dysfunctional cellular responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM7SF3
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TM7SF3
Gene Accession No. Q9NS93
Gene Alias 2010003B14Rik; NARP1; Seven span transmembrane protein; seven transmembrane protein TM7SF3; TM7SF3; Transmembrane 7 superfamily member 3
Gene Symbols TM7SF3
Host Species Rabbit
Immunogen E.coli-derived human TM7SF3 recombinant protein (Position: R102-R544).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51768
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.